Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

VWR® Sharps Container Systems, Red

8 gal., Stackable, 28.6W×28.6D×34.9H cm (11.25W×11.25D×14"H), Red
  19001-010
VWR® Sharps Container Systems, Red
  19001-010
VWR® Sharps Container Systems, Red
VWR® Sharps Container Systems, Red

8 gal., Stackable, 28.6W×28.6D×34.9H cm (11.25W×11.25D×14"H), Red

 
$394.25
   
Each (1UN) Case de 10 (1UN) Paleta de 90 (1UN)
 

Sharps container systems provide solutions for the safe and effective containment and disposal of contaminated needles, syringes, and other sharps. Choose from three different systems in a variety of sizes.

  • Puncture-resistant
  • Autoclave-safe and incinerator-safe
  • Transparent, locking lids
  • Needle removal ports

The extended neck sharps containers feature a funneled vertical drop and extended handles. Puncture-resistant and autoclave- and incinerator-safe.

 
  • Capacity:
    8 gal.
  • Color:
    Red
  • Dimensions:
    28.6W×28.6D×34.9H cm (11.25W×11.25D×14"H)
  • Style:
    Stackable