VWR® Sharps Container Systems, Red
2 gal., Stackable, 25.4W×17.8D×24.1H cm (10W×7D×9.5"H), Red
Supplier: VWR International
Jump to:
Product Details & Documents
Sharps container systems provide solutions for the safe and effective containment and disposal of contaminated needles, syringes, and other sharps. Choose from three different systems in a variety of sizes.
- Puncture-resistant
- Autoclave-safe and incinerator-safe
- Transparent, locking lids
- Needle removal ports
The extended neck sharps containers feature a funneled vertical drop and extended handles. Puncture-resistant and autoclave- and incinerator-safe.
Documents
Especificaciones
- Capacity:2 gal.
- Color:Red
- Dimensions:25.4W×17.8D×24.1H cm (10W×7D×9.5"H)
- Style:Stackable