VWR® Sharps Container Systems, Red
1 qt., Stackable, 8.9W×8.9D×17.8H cm (3.5W×3.5D×7"H), Red
Supplier: VWR International
Jump to:
Product Details & Documents
Sharps container systems provide solutions for the safe and effective containment and disposal of contaminated needles, syringes, and other sharps. Choose from three different systems in a variety of sizes.
- Puncture-resistant
- Autoclave-safe and incinerator-safe
- Transparent, locking lids
- Needle removal ports
The extended neck sharps containers feature a funneled vertical drop and extended handles. Puncture-resistant and autoclave- and incinerator-safe.
Documents
Especificaciones
- Capacity:1 qt.
- Color:Red
- Dimensions:8.9W×8.9D×17.8H cm (3.5W×3.5D×7"H)
- Style:Stackable
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



