Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Flask Brushes, Justman Brush

For 50 mL flasks, Black, 44 - 54 mm, Wire
  17145-052
 :  J620370
Flask Brushes, Justman Brush
  17145-052
Flask Brushes, Justman Brush

For 50 mL flasks, Black, 44 - 54 mm, Wire

 
$135.80
   
Each (1UN) Paquete de 12 (1UN)
 

These brushes with special construction puts the cleaning action directly where it is needed.

The bristles fold flat to fit throught the narrow bottle neck, then expand to fit the chamber.

 
  • Bristle Color:
    Black
  • Brush head Ø:
    44 - 54 mm
  • Handle Material:
    Wire
  • Description:
    For 50 mL flasks