Order Entry
Mexico
Orders LinkContactUsLinkComponent
Flask Brushes, Justman Brush

Flask Brushes, Justman Brush

For 25 mL flasks, Black, 44 mm (1 3/4"), Wire
Catalog # 17145-050
Número de proveedor #:  J620368
Flask Brushes, Justman Brush
Catalog # 17145-050
Flask Brushes, Justman Brush

For 25 mL flasks, Black, 44 mm (1 3/4"), Wire

Price
$200.36
Log in Para disponibilidad
Each (1UN) Paquete de 12 (1UN)
Product Details & Documents

These brushes with special construction puts the cleaning action directly where it is needed.

The bristles fold flat to fit throught the narrow bottle neck, then expand to fit the chamber.

Especificaciones
  • Bristle Color:
    Black
  • Brush head Ø:
    44 mm (1 3/4")
  • Handle Material:
    Wire
  • Description:
    For 25 mL flasks