Straight Plug Rubber Stoppers, Wheaton
Mouth with ID×OD 15×30 mm, Straight plug rubber stoppers, durameter 40, Natural red rubber
Jump to:
Discontinued
Product Details & Documents
Wheaton straight plug stoppers provide maximum sealing for vials with a straight-wall glass finish
Stoppers are autoclavable.
Stoppers are available in a variety of sizes and rubber formulations . Straight plug stoppers are manufactured from the highest quality raw materials and are manufactured to yield exceptional dimensional stability.
Documents
Specifications
- Description:Straight plug rubber stoppers, durameter 40
- Material:Natural red rubber
- Fits:Mouth with ID×OD 15×30 mm