Order Entry
Mexico
Orders LinkContactUsLinkComponent
Sample Vials and Caps, MicroLiter, DWK Life Sciences

Sample Vials and Caps, MicroLiter, DWK Life Sciences

Vial, 4.00 ml, 15×45 mm, Clear, No, 13-425
  97047-630
Supplier #:  13-1000
Sample Vials and Caps, MicroLiter, DWK Life Sciences
  97047-630
Sample Vials and Caps, MicroLiter, DWK Life Sciences
Sample Vials and Caps, MicroLiter, DWK Life Sciences

Vial, 4.00 ml, 15×45 mm, Clear, No, 13-425

 
$816.00
   
Paquete de 100
 

All vials may appear to be created equal, but quality and performance make the difference when analyzing vital samples.

MicroLiter’s product line of sample vials offer superior storage and handling of larger volume samples. Available in clear and amber formats, with polypropylene or phenolic closures and a variety of septa and liner styles.

MicroLiter vials are cleaned and packaged in a certified Class 10000 clean room according to a designed process, developed to eliminate chromatographic errors associated with vials. Critical physical dimensions are fully tested, as well as chemical cleanliness testing to ensure that trace levels of residues are not present. The result of this process are identical, clean, dependable vials, certified to ensure optimal performance and reproducible results.

Certifications: Certified Clean

 
  • Cap included:
    No
  • Cap Size:
    13-425
  • Capacity:
    4.00 ml
  • Color:
    Clear
  • Description:
    Vial
  • Diameter ×H:
    15×45 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.