Orders LinkContactUsLinkComponent
Tax-Fre® Dry Mops, Contec®
Tax-Fre® Dry Mops, Contec®
Tax-Fre® Tack Cloth, Quarterfolded, 25.4×43.2 cm (10×17"), 50/Bag, 8 Bags/Case
Catalog # 94052-810
Tax-Fre® Dry Mops, Contec®
Catalog # 94052-810
Supplier:  
Tax-Fre® Dry Mops, Contec®
Tax-Fre® Dry Mops, Contec®
Tax-Fre® Tack Cloth, Quarterfolded, 25.4×43.2 cm (10×17"), 50/Bag, 8 Bags/Case
Discontinued
Product Details & Documents
Designed for dry mopping floors and walls, the Tax-Fre® Dry Mop utilizes the same low-residue material as Tax-Fre® wipes.
  • Utilizing a low-residue pressure-sensitive material, this dry floor mop provides superior pick-up in areas where wet mopping is not an option
  • Will not scratch or damage sensitive surfaces
  • Dramatically reduces clean-up time
  • Ideal for use when liquids are not recommended
The 23×5" (58.4×12.7 cm) trapezoid-shaped swivel head attached to a lightweight 5' (1.5 m) handle enables the operator to clean in hard to each areas. All hardware is cleanroom compatible. The unique construction of the Tax-Fre® mop head allows the use of multiple sheets that can easily be removed when contaminated, revealing a fresh sheet for continued cleaning.
Especificaciones
  • Description:
    Tax-Fre® Tack Cloth, Quarterfolded
  • Dimensions:
    25.4×43.2 cm (10×17")
  • Packed:
    50/Bag, 8 Bags/Case