Tax-Fre® Dry Mops, Contec®
Tax-Fre® Tack Cloth, Quarterfolded, 25.4×43.2 cm (10×17"), 50/Bag, 8 Bags/Case
Jump to:
Discontinued
Product Details & Documents
Designed for dry mopping floors and walls, the Tax-Fre® Dry Mop utilizes the same low-residue material as Tax-Fre® wipes.
- Utilizing a low-residue pressure-sensitive material, this dry floor mop provides superior pick-up in areas where wet mopping is not an option
- Will not scratch or damage sensitive surfaces
- Dramatically reduces clean-up time
- Ideal for use when liquids are not recommended
The 23×5" (58.4×12.7 cm) trapezoid-shaped swivel head attached to a lightweight 5' (1.5 m) handle enables the operator to clean in hard to each areas. All hardware is cleanroom compatible. The unique construction of the Tax-Fre® mop head allows the use of multiple sheets that can easily be removed when contaminated, revealing a fresh sheet for continued cleaning.
Documents
Especificaciones
- Description:Tax-Fre® Tack Cloth, Quarterfolded
- Dimensions:25.4×43.2 cm (10×17")
- Packed:50/Bag, 8 Bags/Case