Wringers, Contec®
Downpress Flat Mop Wringer, Stainless steel, VertiKlean® MAX™ and QuickTask™ Mops
Supplier: CONTEC, INC. MX
Número de proveedor #: 2756
Jump to:
Product Details & Documents
A variety of wringers designed for use with curved and flat mops.
- Stainless steel and polypropylene wringers for a variety of Contec® bucket systems
- Can be used with curved, flat, and string mops
- Fully autoclavable
These wringers fit on Contec® 3.9 gallon (15 l), 5 gallon (20 l) , and 6.5 gallon (25 l) bucket systems. All wringers are compatible with a wide variety of disinfectants.
Especificaciones
- For Use With:VertiKlean® MAX™ and QuickTask™ Mops
- Description:Downpress Flat Mop Wringer
- Material:Stainless steel