Orders LinkContactUsLinkComponent
PYREX® Separatory Funnels, Squibb, Pear-Shaped, Corning
60 ml, 2, 16
  89091-514
Product available on GSA Advantage®
   Low Manufacturing Impact
 :   Corning
 :  6402-60
PYREX® Separatory Funnels, Squibb, Pear-Shaped, Corning
  89091-514
 :  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
PYREX® Separatory Funnels, Squibb, Pear-Shaped, Corning
PYREX® Separatory Funnels, Squibb, Pear-Shaped, Corning
60 ml, 2, 16
$17,530.00
Case (4UN)
PYREX® separatory funnels, squibb, pear-shaped, Corning
Reduces freezing problems and grease contamination.
  • Capacity:
    60 ml
  • ST Stopper No.:
    16
  • Stopcock Size:
    2
  • Cat. No.:
    89091-514
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Environmentally Preferable:
    Low Manufacturing Impact