Orders LinkContactUsLinkComponent
PYREX® Round-Bottom Boiling Flasks, Short Ring Neck, Corning
2 L, 10, 162 x 232 mm (63/8 x 91/8")
Catalog # 89091-296
Supplier:   Corning
PYREX® Round-Bottom Boiling Flasks, Short Ring Neck, Corning
Catalog # 89091-296
Supplier:  Corning
PYREX® Round-Bottom Boiling Flasks, Short Ring Neck, Corning
PYREX® Round-Bottom Boiling Flasks, Short Ring Neck, Corning
2 L, 10, 162 x 232 mm (63/8 x 91/8")
Price
$36,390.00
Log in Para disponibilidad
Case (18UN)
Product Details & Documents
PYREX® borosilicate glass, clear, with short neck.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These flask mouth tooled for uniform rubber stopper fit. The normal working temperature is 230 °C.
Especificaciones
  • Capacity:
    2 L
  • Stopper No.:
    10
  • O.D.×H:
    162 x 232 mm (63/8 x 91/8")