Orders LinkContactUsLinkComponent
Volumetric Pipettes, Class A, Color-Coded, Hirschmann
Unserialized Pipets, 1 ml, Blue, ±0.006 ml
Catalog # 89085-152
Supplier:   HIRSCHMANN INC
 :  134G1
Volumetric Pipettes, Class A, Color-Coded, Hirschmann
Catalog # 89085-152
Supplier:  HIRSCHMANN INC
Volumetric Pipettes, Class A, Color-Coded, Hirschmann
Volumetric Pipettes, Class A, Color-Coded, Hirschmann
Unserialized Pipets, 1 ml, Blue, ±0.006 ml
Price
$5,303.07
Log in Para disponibilidad
Paquete de 12
Product Details & Documents
Pipettes are constructed of borosilicate glass and feature black enamel lettering and a graduation line.
Designed to deliver according to ASTM Specification E969, Class A requirements. Color-coded for ease in sorting and selection according to ASTM E1273. Serialized pipettes include a Certificate of Identification and Accuracy. All pipettes feature lot number and production year printed onto the pipette for QA purposes.
Especificaciones
  • Capacity:
    1 ml
  • Description:
    Unserialized Pipets
  • Tolerance:
    ±0.006 ml
  • Band Color:
    Blue