Orders LinkContactUsLinkComponent
VWR® Thin Sleeves, PTFE
VWR® Thin Sleeves, PTFE
24/40
Catalog # 89026-488
Supplier:   VWR International
VWR® Thin Sleeves, PTFE
Catalog # 89026-488
Supplier:  VWR International
VWR® Thin Sleeves, PTFE
VWR® Thin Sleeves, PTFE
24/40
Price
$690.09
Log in Para disponibilidad
Paquete de 10
Product Details & Documents
These PTFE sleeves withstand low to medium pressure and are inserted between standard ground glass joints to eliminate the use of grease and prevent sticking.
  • Chemically resistant
  • Inert
  • PTFE fabrication
Sleeves are inert, have a wall thickness of about 0.05mm, and are usable to 280°C (536°F).
Especificaciones
  • ST Joint No.:
    24/40