Orders LinkContactUsLinkComponent
VWR®, Beakers, PTFE
5 L (169 oz.), 280 mm, 170 mm
  89026-028
VWR®, Beakers, PTFE
  89026-028
VWR®, Beakers, PTFE
VWR®, Beakers, PTFE
5 L (169 oz.), 280 mm, 170 mm
$15,195.52
Each (1UN)
Isostatically molded in pure, FDA approved PTFE, these beakers feature a pouring spout and have a machined base for good heat transfer.
  • Chemically resistant and inert
  • Autoclavable
Machined base provides good heat transfer in these PTFE beakers.
  • Ext.Ø:
    170 mm
  • Capacity:
    5 L (169 oz.)
  • Height:
    280 mm
Accessories
  •  
    Description
    For Use With
  • 89026-434
    Beaker Covers
    100 mL Beaker
    Paquete de 2 - $553.26