VWR® HDPE Multipurpose Containers
946 ml (32 oz.), 11.4 dia.×12H cm (4¹/₂×4³/₄")
Supplier: VWR International
Jump to:
Product Details & Documents
Translucent containers for the storage of pathology, industrial, and environmental samples. Containers are made of high quality plastic to resist breakage and come in a wide range of sizes. The matching cap snaps on tight for a secure fit. The 4800mL container is equipped with a handle for portability.
- HDPE construction
- Chemical-resistant
- Translucent siding
Containers are made of high-quality plastic to resist breakage and come in a wide range of sizes. The matching cap snaps on tight for a secure fit. The 4800 mL container is equipped with a handle for portability.
Documents
Certificates
Buscar por número de lote89009-662_P 89009-664_P 89009-666_P 89009-668_P 89009-670_P
BSE/TSE Certificates
More
Especificaciones
- Capacity:946 ml (32 oz.)
- Dimensions:11.4 dia.×12H cm (4¹/₂×4³/₄")