Corning® Coliform Water Test Sample Container, Corning
Coliform water test sample container with Thiosulfate tablet, 120.00 ml, Yes
Supplier: Corning
Número de proveedor #: 1700-100
Jump to:
Product Details & Documents
These graduated, sterile, cylindrical vials made from PP are used in testing for the presence of coliform bacteria, microbiological contaminants in drinking water.
- One-piece container with attached lid to reduce chance of contamination
- Locking arrow ensures sterility
- Leak tight when sealed properly
- Tie-down to protect from accidental opening
- With or without sodium thiosulfate tablet
Meets EPA requirements
Ordering information: Includes 100 containers per case. Comes with sample labels and instructions.
Especificaciones
- Cap included:Yes
- Capacity:120.00 ml
- Description:Coliform water test sample container with Thiosulfate tablet