Order Entry
Mexico
Orders LinkContactUsLinkComponent
Sample Vials, Clear Borosilicate Glass, Screw-Thread

Sample Vials, Clear Borosilicate Glass, Screw-Thread

Vials without treatment, 1.90 ml, 12×35 mm, Clear, Yes, 8-425
Catalog # 66009-556
Supplier:   QORPAK
Supplier #:  GLC-00978
Sample Vials, Clear Borosilicate Glass, Screw-Thread
Catalog # 66009-556
Supplier:  QORPAK
Sample Vials, Clear Borosilicate Glass, Screw-Thread
Sample Vials, Clear Borosilicate Glass, Screw-Thread

Vials without treatment, 1.90 ml, 12×35 mm, Clear, Yes, 8-425

Price
$4,244.24
Log in Para disponibilidad
Case (144UN)
Product Details & Documents

Clear, flint screw-thread sample vials offer maximum visibility.

  • Available with a variety of caps, or as vials only, to meet specific requirements
  • Pre-cleaned options save time and money

These general-purpose glass vials are made of Type I Borosilicate glass, providing exceptional resistance to heat shock and chemical leaching.

Especificaciones
  • Cap included:
    Yes
  • Cap Size:
    8-425
  • Capacity:
    1.90 ml
  • Color:
    Clear
  • Description:
    Vials without treatment
  • Diameter ×H:
    12×35 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.