Accessories for Brushes, Scratch, Replaceable Tips, Excelta Corp®
Replacement tip for 267, 2 star quality, Fiberglass, 0.13" (3.25 mm)
Catalog # 63041-740
Supplier: EXCELTA CORPORATION
Número de proveedor #: 267A
Accessories for Brushes, Scratch, Replaceable Tips, Excelta Corp®
Catalog # 63041-740
Supplier: EXCELTA CORPORATION
Jump to:
Product Details & Documents
Especificaciones
- Bristle Material:Fiberglass
- Description:Replacement tip for 267, 2 star quality
- Brush Head Width:0.13" (3.25 mm)