Accessories for Brushes, Scratch, Replaceable Tips, Excelta Corp®
Replacement tip for 265, 2 star quality, Brass, 0.13" (3.25 mm)
Catalog # 63041-732
Supplier: EXCELTA CORPORATION
Supplier #: 265A
Accessories for Brushes, Scratch, Replaceable Tips, Excelta Corp®
Catalog # 63041-732
Supplier: EXCELTA CORPORATION
Jump to:
Product Details & Documents
Specifications
- Bristle Material:Brass
- Description:Replacement tip for 265, 2 star quality
- Brush Head Width:0.13" (3.25 mm)