pH Test Papers, Precision Laboratories
Potassium Iodide Starch, 100 strips/pack, Vial
Catalog # 60799-008
Supplier: PRECISION LABORATORIES
Supplier #: VWR-160-25V
Jump to:
Product Details & Documents
Available in 8 books of 25 sheets, or vials containing 100 sheets.
Specifications
- Description:Potassium Iodide Starch
- Size:100 strips/pack
- Packaging:Vial