[ST] Joint Clips
Plastic joint clip, 24/40
Catalog # 60004-950
Supplier: KEMTECH AMERICA INC
Número de proveedor #: CS002440
Jump to:
Product Details & Documents
Made of plastic.
- Heavy duty plastic construction
- Color coded
- Fit standard taper joints
Our heavy duty keck clips are perfect for all standard taper ground glass joints.
Especificaciones
- Description:Plastic joint clip
- ST Joint Size:24/40