Hard Rubber Spatulas
152 mm (6")
57930-047
: H/R-6"
Ideal for use with materials that affect metals adversely, for mixing, weighing, cleaning, blending, and many other uses.
- Non sparking
- Wide and flexible blade
Hand-finished and hand-polished blades. Spatulas are rust-proof, washable, and sterilizable.
- Blade Length:152 mm (6")