Orders LinkContactUsLinkComponent
Hard Rubber Spatulas
152 mm (6")
  57930-047
 :  H/R-6"
Hard Rubber Spatulas
  57930-047
Hard Rubber Spatulas
Hard Rubber Spatulas
152 mm (6")
$514.40
Each (1UN) Paquete de 12 (1UN)
Ideal for use with materials that affect metals adversely, for mixing, weighing, cleaning, blending, and many other uses.
  • Non sparking
  • Wide and flexible blade
Hand-finished and hand-polished blades. Spatulas are rust-proof, washable, and sterilizable.
  • Blade Length:
    152 mm (6")