Hard Rubber Spatulas
102 mm (4")
Catalog # 57930-025
Supplier: JOHN M. MARIS CO
: H/R-4"
Jump to:
Product Details & Documents
Ideal for use with materials that affect metals adversely, for mixing, weighing, cleaning, blending, and many other uses.
- Non sparking
- Wide and flexible blade
Hand-finished and hand-polished blades. Spatulas are rust-proof, washable, and sterilizable.
Especificaciones
- Blade Length:102 mm (4")