Orders LinkContactUsLinkComponent
Hard Rubber Spatulas
102 mm (4")
Catalog # 57930-025
Supplier:   JOHN M. MARIS CO
 :  H/R-4"
Hard Rubber Spatulas
Catalog # 57930-025
Supplier:  JOHN M. MARIS CO
Hard Rubber Spatulas
Hard Rubber Spatulas
102 mm (4")
Price
$467.98
Log in Para disponibilidad
Each (1UN) Paquete de 12 (1UN)
Product Details & Documents
Ideal for use with materials that affect metals adversely, for mixing, weighing, cleaning, blending, and many other uses.
  • Non sparking
  • Wide and flexible blade
Hand-finished and hand-polished blades. Spatulas are rust-proof, washable, and sterilizable.
Especificaciones
  • Blade Length:
    102 mm (4")