Orders LinkContactUsLinkComponent
Accessories for Sonic Sifter Separator, Advantech
Fines Collector for Sonic Sifter Separator
  57334-040
 :  L3-N5
Accessories for Sonic Sifter Separator, Advantech
  57334-040
Accessories for Sonic Sifter Separator, Advantech
Accessories for Sonic Sifter Separator, Advantech
Fines Collector for Sonic Sifter Separator
$2,489.05
Each (1UN)
  • Description:
    Fines Collector for Sonic Sifter Separator
Accessories
  •  
    Height
    Mesh Size
    USA Series
  • 57334-010
    15.9 mm (⅝")
    250 µm (0.0098")
    #60
    Each (1UN) - $4,346.74
  •  
    Height
    Mesh Size
    USA Series
  • 57334-016
    15.9 mm (⅝")
    150 µm (0.0059")
    #100
    Each (1UN) - $4,546.11
  •  
    Height
    Mesh Size
    USA Series
  • 57334-020
    15.9 mm (⅝")
    106 µm (0.0042")
    #140
    Each (1UN) - $4,546.11
  •  
    Height
    Mesh Size
    USA Series
  • 57334-022
    15.9 mm (⅝")
    90 µm (0.0035")
    #170
    Each (1UN) - $4,546.11
  •  
    Height
    Mesh Size
    USA Series
  • 57334-024
    15.9 mm (⅝")
    75 µm (0.0030")
    #200
    Each (1UN) - $4,945.31
  •  
    Height
    Mesh Size
    USA Series
  • 57334-030
    15.9 mm (⅝")
    45 µm (0.0018")
    #325
    Each (1UN) - $8,242.17
  •  
    Description
  • 57334-034
    Diaphragm for Sonic Sifter Separator
    Each (1UN) - $1,778.41
  •  
    Height
    Mesh Size
    USA Series
  • 57333-774
    15.9 mm (⅝")
    710 µm (0.0280")
    #25
    Each (1UN) - $4,346.74
  •  
    Height
    Mesh Size
    USA Series
  • 57333-770
    15.9 mm (⅝")
    1.18 mm (0.0465")
    #16
    Each (1UN) - $4,346.74
  •  
    Description
  • 57334-040
    Fines Collector for Sonic Sifter Separator
    Each (1UN) - $2,489.05
  •  
    Height
    Mesh Size
    USA Series
  • 57334-004
    15.9 mm (⅝")
    425 µm (0.0167")
    #40
    Each (1UN) - $4,346.74
  •  
    Height
    Mesh Size
    USA Series
  • 57334-002
    15.9 mm (⅝")
    600 µm (0.0236")
    #30
    Each (1UN) - $4,346.74
  •  
    Height
    Mesh Size
    USA Series
  • 57334-000
    15.9 mm (⅝")
    850 µm (0.0334")
    #20
    Each (1UN) - $4,346.74