VWR® Microdispenser Replacement Kits
2500 uL VWR® Microdispenser Replacement Kit
Catalog # 53506-928
Supplier: VWR International
Jump to:
Product Details & Documents
The 5, 10, and 20 µl VWR® Microdispenser Replacement Kits each include 2 plungers, 2 O-rings, 1 collet, and 1 coil spring and barrel tube.
- Available in a variety of sizes
Delivery information: The 20 and 25, 50, 100, 100/200, and 250 µl VWR® Microdispenser Replacement Kits each include 1 plunger, 2 O-rings, 1 collet, 3 fluoropolymer resin tips, and 1 coil spring and barrel tube.
Specifications
- Description:2500 uL VWR® Microdispenser Replacement Kit