VWR® Lab Markers
Lab marker #2, Blue, Fine
Supplier: VWR International
Jump to:
Product Details & Documents
Special-purpose markers write through moisture as easily as they write on dry surfaces, and are suitable for industrial, biomedical, and educational laboratories.
- Provide permanent marking on plastic disposables
- Dries quickly for immediate handling of labware
- Durable tip and generous ink supply provide a long product life
Markers provide permanent marking on plastic disposables. When used on glass or metal reusables, or on porcelain surfaces, ink will not smear when subjected to water and can be washed off these surfaces using various solvents, allowing repeated marking.
Especificaciones
- Point Size:Fine
- Color:Blue
- Description:Lab marker #2