Orders LinkContactUsLinkComponent
Accessories for Single-Sample Osmometer, Model 3250, Advanced Instruments
Accessories for Single-Sample Osmometer, Model 3250, Advanced Instruments
500 mOsm Calibration Standard, 5 mL
Catalog # 52197-445
Número de proveedor #:  3LA051
Accessories for Single-Sample Osmometer, Model 3250, Advanced Instruments
Catalog # 52197-445
Accessories for Single-Sample Osmometer, Model 3250, Advanced Instruments
500 mOsm Calibration Standard, 5 mL
Price
$7,972.80
Log in Para disponibilidad
Paquete de 10 (5ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Documents
Especificaciones
  • Description:
    500 mOsm Calibration Standard, 5 mL
Accessories
Información del producto
  • Catalog #
    Availability
    Price
  • Sample Tubes, 0.2 mL
    Sample Tubes, 0.2 mL
    52197-167
    Log in Para disponibilidad
    Paquete de 500 - $64,870.50
Información del producto
  • Catalog #
    Availability
    Price
  • Clinitrol™ 290 Reference Solution, 5 mL Ampules
    Clinitrol™ 290 Reference Solution, 5 mL Ampules
    52197-076
    Log in Para disponibilidad
    Paquete de 10 - $9,784.64
Información del producto
  • Catalog #
    Availability
    Price
  • Osmolality Linearity Set, 5 Level, 10 x 5 mL
    Osmolality Linearity Set, 5 Level, 10 x 5 mL
    52198-164
    Log in Para disponibilidad
    Each (1UN) - $9,462.10
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 