Orders LinkContactUsLinkComponent
Accessories for Multi-Sample Osmometer, Model 3900, Advanced Instruments
Accessories for Multi-Sample Osmometer, Model 3900, Advanced Instruments
Sample Tubes, 0.2 mL
Catalog # 52197-167
Número de proveedor #:  3LA825
Accessories for Multi-Sample Osmometer, Model 3900, Advanced Instruments
Catalog # 52197-167
Accessories for Multi-Sample Osmometer, Model 3900, Advanced Instruments
Sample Tubes, 0.2 mL
Price
$64,870.50
Log in Para disponibilidad
Paquete de 500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Especificaciones
  • Description:
    Sample Tubes, 0.2 mL
Accessories
Información del producto
  • Catalog #
    Availability
    Price
  • 100mOsm Calibration Standard, 5 mL
    100mOsm Calibration Standard, 5 mL
    52197-442
    Log in Para disponibilidad
    Paquete de 10 (5ml) - $7,202.77
Información del producto
  • Catalog #
    Availability
    Price
  • 500 mOsm Calibration Standard, 5 mL
    500 mOsm Calibration Standard, 5 mL
    52197-445
    Log in Para disponibilidad
    Paquete de 10 (5ml) - $7,972.80
Información del producto
  • Catalog #
    Availability
    Price
  • Osmolality Linearity Set, 5 Level, 10 x 5 mL
    Osmolality Linearity Set, 5 Level, 10 x 5 mL
    52198-164
    Log in Para disponibilidad
    Each (1UN) - $9,462.10
Información del producto
  • Catalog #
    Availability
    Price
  • Heat Transfer Fluid, 150 mL
    Heat Transfer Fluid, 150 mL
    52197-434
    Log in Para disponibilidad
    Each (150ml) - $4,069.21
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 