Orders LinkContactUsLinkComponent
Orion™ pH Buffer Bottles, Thermo Scientific
Orion™ pH Buffer Bottles, Thermo Scientific
pH 7.00 buffer, 475 ml, Yellow
Catalog # 34182-386
Supplier #:  910107
Orion™ pH Buffer Bottles, Thermo Scientific
Catalog # 34182-386
CAS Number:  
Orion™ pH Buffer Bottles, Thermo Scientific
Orion™ pH Buffer Bottles, Thermo Scientific
pH 7.00 buffer, 475 ml, Yellow
CAS Number:  
Price
$740.00
Log in for availability
Each (475ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Perform accurate and reproducible pH calibrations with Orion™ pH buffer bottles.
  • All bottles feature pH/temperature tables printed on the labels for accurate calibrations and NIST traceability
  • Manufactured under ISO 9000 quality standards
  • Available in pint (475 ml) bottles or portable five packs of 2 oz. (5×60 ml) bottles
The pH 4.01, 5.00, 7.00, and 10.01 buffers are color-coded for ease of use. All Orion™ pH buffers are traceable to NIST standard reference materials.
Documents
Specifications
Specifications
  • Color:
    Yellow
  • Description:
    pH 7.00 buffer
  • Size:
    475 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.