Order Entry
Mexico
Orders LinkContactUsLinkComponent
Orion™ pH Buffer Bottles, Thermo Scientific

Orion™ pH Buffer Bottles, Thermo Scientific

pH 5.00 buffer, 475 ml, Orange
Catalog # 34182-310
Supplier #:  910105
Orion™ pH Buffer Bottles, Thermo Scientific
Catalog # 34182-310
Supplier:  
CAS Number:  
Orion™ pH Buffer Bottles, Thermo Scientific
Orion™ pH Buffer Bottles, Thermo Scientific

pH 5.00 buffer, 475 ml, Orange

CAS Number:  
Discontinued
El producto ha sido descontinuado. Sin embargo, encontramos productos alternativos similares.
Alternativas
BDH BUFFER REF STD PH5 COLORLESS 500ML Catalog #BDH5034-500ML

Product BDH5034-500ML is similar to 34182-310 but manufacturer and specifications may vary.

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Perform accurate and reproducible pH calibrations with Orion™ pH buffer bottles.

  • All bottles feature pH/temperature tables printed on the labels for accurate calibrations and NIST traceability
  • Manufactured under ISO 9000 quality standards
  • Available in pint (475 ml) bottles or portable five packs of 2 oz. (5×60 ml) bottles

The pH 4.01, 5.00, 7.00, and 10.01 buffers are color-coded for ease of use. All Orion™ pH buffers are traceable to NIST standard reference materials.

Documents

Especificaciones

Especificaciones

  • Color:
    Orange
  • Description:
    pH 5.00 buffer
  • Size:
    475 ml

Especificaciones

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.