Order Entry
Mexico
Orders LinkContactUsLinkComponent
Zero Pressure Weather Caps, Labconco®

Zero Pressure Weather Caps, Labconco®

Zero Pressure Weather Caps, 142.2 cm (56"), 30.5 cm (12"), 15.9 kg (35 lbs.)
Catalog # 30185-000
Supplier #:  5622100
Zero Pressure Weather Caps, Labconco®
Catalog # 30185-000
Zero Pressure Weather Caps, Labconco®
Zero Pressure Weather Caps, Labconco®

Zero Pressure Weather Caps, 142.2 cm (56"), 30.5 cm (12"), 15.9 kg (35 lbs.)

Price
$32,570.00
Log in for availability
Each (1UN)
Product Details & Documents

Weathercaps are constructed of strong, corrosion-resistant PVC.

Caps fits atop standard PVC duct, and permit vertical discharge of effluent air above roofline for dispersion away from the building. Caps add little static pressure to the exhaust system.

Specifications
  • Description:
    Zero Pressure Weather Caps
  • Height:
    142.2 cm (56")
  • Shipping Weight:
    15.9 kg (35 lbs.)
  • Nominal Diameter:
    30.5 cm (12")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.