Universal Fit Pipet Tips, Corning®
1 - 200 µl, -, Yellow, Rack Packed
Supplier: Corning
: 4865
Universal Fit Pipet Tips, Corning®
Catalog # 29442-686
Supplier: Corning
Product available on GSA Advantage®
Jump to:
Product Details & Documents
Designed to assure a reliable fit with all major brand pipettors. Beveled tip ensures full sample delivery. All tips are nonpyrogenic. All 1–200µL rack packed tips are RNase- and DNase-free.
- Beveled orifice helps ensure accurate fluid delivery
- All tips are certified RNase-/DNase-free and non pyrogenic
- Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design
- Bulk packed tips are non sterile and very economical
1 to 200 µl universal fit tips are graduated at the 10 µl, 50 µl, and 100 µl volumes.
Documents
Certificates
Buscar por número de loteCorning® Aspirator A Convenient User-friendly device that facilates the aspiration of Liquids From Lab Containers Corning® ELISA Microplates for Biochemical and Cell-based Applications Pall Laboratory Consumables Reagents and Buffers for Genomic DNA Purification Pall Laboratory Standard Reagent Protocol for Genomic DNA Isolation
Product Literature
Especificaciones
- Volume:1 - 200 µl
- Tip Color:Yellow
- Packed:Rack Packed
- Sterile:-
- Cat. No.:29442-686
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Environmentally Preferable:Low Manufacturing Impact