Orders LinkContactUsLinkComponent
Universal Fit Pipet Tips, Corning®
1 - 200 µl, -, Yellow, Rack Packed
Catalog # 29442-686
Product available on GSA Advantage®
   Low Manufacturing Impact
Supplier:   Corning
 :  4865
Universal Fit Pipet Tips, Corning®
Catalog # 29442-686
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
Universal Fit Pipet Tips, Corning®
Universal Fit Pipet Tips, Corning®
1 - 200 µl, -, Yellow, Rack Packed
Price
$1,310.00
Log in Para disponibilidad
Case (960UN)
Product Details & Documents
Designed to assure a reliable fit with all major brand pipettors. Beveled tip ensures full sample delivery. All tips are nonpyrogenic. All 1–200µL rack packed tips are RNase- and DNase-free.
  • Beveled orifice helps ensure accurate fluid delivery
  • All tips are certified RNase-/DNase-free and non pyrogenic
  • Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design
  • Bulk packed tips are non sterile and very economical
1 to 200 µl universal fit tips are graduated at the 10 µl, 50 µl, and 100 µl volumes.
Especificaciones
  • Volume:
    1 - 200 µl
  • Tip Color:
    Yellow
  • Packed:
    Rack Packed
  • Sterile:
    -
  • Cat. No.:
    29442-686
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Environmentally Preferable:
    Low Manufacturing Impact