96-Well Polypropylene Tubes, Corning®
1.2 ml, 8-Tube Strips, 12/Rack, Sterile
Supplier: Corning
Número de proveedor #: 4413
Jump to:
Product Details & Documents
Cluster tube system is composed of ninety-six 1.2mL polypropylene tubes arranged in a microtitration plate format. Tubes are available individually or in strips of eight tubes.
- Composed of 96 PP tubes in a standard microplate format
- 1.2 ml tubes available individually or in strips of 8 tubes
- PE tube caps available in 8-cap strips
Packaging: Supplied bagged or racked.
Especificaciones
- Sterility:Sterile
- Capacity:1.2 ml
- Description:8-Tube Strips
- Packaging:12/Rack



