Orders LinkContactUsLinkComponent
Whatman™ 1PS Phase Separator Papers
Whatman™ 1PS Phase Separator Papers
1PS Phase Separator for Solvent Extraction, 125 mm circle, 125 mm
Catalog # 28491-146
Supplier:   Cytiva
Número de proveedor #:  2200-125
Whatman™ 1PS Phase Separator Papers
Catalog # 28491-146
Supplier:  Cytiva
Whatman™ 1PS Phase Separator Papers
Whatman™ 1PS Phase Separator Papers
1PS Phase Separator for Solvent Extraction, 125 mm circle, 125 mm
Price
$3,933.51
Log in Para disponibilidad
Paquete de 100
Product Details & Documents
Whatman 1PS Phase Separator from Cytiva's business is a hydrophobic filter for routine solvent extraction. Within seconds it retains aqueous phases, while solvent phases flow through.
  • Impregnated with stabilized silicone to create a hydrophobic filter
  • Multiple separations can be processed in parallel
  • No special training required
  • Can replace a separatory funnel in certain applications
Solvent extraction can be time-consuming and messy when numerous separations are required
Mixed phases are poured directly into the Whatman 1PS Phase Separator filter. The solvent phase flows through the paper quickly and cleanly, automatically stopping after the process is complete; all of the aqueous phase is retained. Droplets are automatically separated within seconds.
Cytiva's Whatman 1PS Phase Separator is impregnated with stabilized silicone, creating a hydrophobic filter paper that can replace separatory funnel use in certain applications.
Especificaciones
  • Ø:
    125 mm
  • Description:
    1PS Phase Separator for Solvent Extraction, 125 mm circle