Orders LinkContactUsLinkComponent
Zero Pressure Weather Caps, Labconco®
Zero Pressure Weather Caps, 121.9 cm (48"), 25.4 cm (10"), 13.6 kg (30 lbs.)
Catalog # 26674-520
 :  7095100
Zero Pressure Weather Caps, Labconco®
Catalog # 26674-520
Supplier:  
Zero Pressure Weather Caps, Labconco®
Zero Pressure Weather Caps, Labconco®
Zero Pressure Weather Caps, 121.9 cm (48"), 25.4 cm (10"), 13.6 kg (30 lbs.)
Discontinued
Product Details & Documents
Weathercaps are constructed of strong, corrosion-resistant PVC.
Caps fits atop standard PVC duct, and permit vertical discharge of effluent air above roofline for dispersion away from the building. Caps add little static pressure to the exhaust system.
Especificaciones
  • Description:
    Zero Pressure Weather Caps
  • Height:
    121.9 cm (48")
  • Shipping Weight:
    13.6 kg (30 lbs.)
  • Nominal Diameter:
    25.4 cm (10")