Orders LinkContactUsLinkComponent
Pipe Cleaner Brushes, Mill-Rose
5 mm (13/64") diameter, 15 m (50')
  17227-044
 :  MR51809
Pipe Cleaner Brushes, Mill-Rose
  17227-044
Pipe Cleaner Brushes, Mill-Rose
Pipe Cleaner Brushes, Mill-Rose
5 mm (13/64") diameter, 15 m (50')
$639.57
Paquete de 50
Continuous lengths of cotton pipe cleaner can be cut as needed for cleaning apertures and narrow tubing.
  • Galvanized wire base provides a strong flexible core
  • For tubes with bottoms, bending the cut end back on itself will help eliminate scratches
For larger bores, a length can be coiled around a finger, tube, or jar for a quick springy brush.
  • Description:
    5 mm (13/64") diameter
  • Total length:
    15 m (50')