Orders LinkContactUsLinkComponent
Straight Plug Rubber Stoppers, Wheaton
Straight Plug Rubber Stoppers, Wheaton
Mouth with ID×OD 15×30 mm, Straight plug rubber stoppers, durameter 50, Gray bromobutyl
Catalog # 16171-630
Straight Plug Rubber Stoppers, Wheaton
Catalog # 16171-630
Supplier:  
Straight Plug Rubber Stoppers, Wheaton
Straight Plug Rubber Stoppers, Wheaton
Mouth with ID×OD 15×30 mm, Straight plug rubber stoppers, durameter 50, Gray bromobutyl
Discontinued
Product Details & Documents
Wheaton straight plug stoppers provide maximum sealing for vials with a straight-wall glass finish
Stoppers are autoclavable.
Stoppers are available in a variety of sizes and rubber formulations . Straight plug stoppers are manufactured from the highest quality raw materials and are manufactured to yield exceptional dimensional stability.
Especificaciones
  • Description:
    Straight plug rubber stoppers, durameter 50
  • Material:
    Gray bromobutyl
  • Fits:
    Mouth with ID×OD 15×30 mm