Polypropylene Snap-Seal™ Sample Containers, Corning®
110 ml (3.7 oz.), Low-Profile Snap-Seal™ Sample Containers
Supplier: Corning
Número de proveedor #: 1730-4L
Jump to:
Product Details & Documents
Designed for a wide variety of applications, these containers provide a reliable leak-tight seal when closed properly.
- Snap-Seal locking device keeps the cap closed and secure
- Hinged cap reduces the chance of sample contamination
- Graduations in both millilitres and ounces
- Rough source for marking
The 120mL (4oz.) size is available in both high- and low-profile design.
Especificaciones
- Capacity:110 ml (3.7 oz.)
- Description:Low-Profile Snap-Seal™ Sample Containers