Whatman™ Polyester Drain Discs
Polyester drain disc, PE, membrane filter accessories, 10 mm, 100 µm
Supplier: Cytiva
Número de proveedor #: 230300
Jump to:
Product Details & Documents
The polyester drain disc is binder free and has a thickness of 100 µm.
- Flat surface to eliminate filter tearing or rupturing
- Used as a separator between membrane layers in serial stack filtration applications
This chemically inert support disc is available in a variety of diameters for use in a range of devices. With Application in General laboratory microfiltration, Quality control and sterility testing, Removal of particulates from HPLC solvents, Tissue culture media filtration.
Documents
Certificates
Buscar por número de loteCytiva CyDye Licensing Statements Cytiva General Licensing Statements
Detalles adicionales del producto
Especificaciones
- Ø:10 mm
- Description:Polyester drain disc, PE, membrane filter accessories
- Thickness:100 µm