VWR®, Beaker Cups, Polystyrene
150 ml (5 oz.)
Supplier: VWR International
Jump to:
Product Details & Documents
Beaker cups are designed for use with various automated procedures and as an economical, general-purpose sample cup.
- Disposable
- Polystyrene fabrication
- Economical sample cup
The 5 mL and 20 mL sizes are recommended for flame photometry analysis.
Especificaciones
- Capacity:150 ml (5 oz.)