VWR®, Beaker Cups, Polystyrene
20 ml (0.68 oz.)
Catalog # 13916-004
Supplier: VWR International
Jump to:
Product Details & Documents
Beaker cups are designed for use with various automated procedures and as an economical, general-purpose sample cup.
- Disposable
- Polystyrene fabrication
- Economical sample cup
The 5 mL and 20 mL sizes are recommended for flame photometry analysis.
Documents
Certificates
Buscar por número de lote13915-996_P 13916-004_P 13916-015_P 13916-037_P 13916-060_P
Certificados BSE/TSE
Especificaciones
- Capacity:20 ml (0.68 oz.)