VWR® Talon® Support Stands, Cast Iron, Rectangular Base
20.3 x 25.4 cm (8 x 10")
Catalog # 12985-074
Supplier: VWR International
VWR® Talon® Support Stands, Cast Iron, Rectangular Base
Catalog # 12985-074
Supplier: VWR International
Jump to:
Product Details & Documents
Support stand bases are constructed of cast iron with black enamel finish.
- Cast iron construction with black enamel finish
- Built-in support rod holder
- Rectangular base
- For use with VWR® Talon® lab frames
All bases feature a built-in support rod holder with locking knob that accepts a 13mm (1/2") diameter support rod (sold separately).
Documents
VWR Laboratory Equipments
Documentación del producto
Especificaciones
- Dimensions:20.3 x 25.4 cm (8 x 10")
Accessories
Información del producto
- Catalog #ØMaterialAvailabilityPrice
- Support Rod
Información del producto
- Catalog #ØMaterialAvailabilityPrice
- Support Rod