Orders LinkContactUsLinkComponent
VWR® Talon® Support Stands, Cast Iron, Rectangular Base
VWR® Talon® Support Stands, Cast Iron, Rectangular Base
20.3 x 25.4 cm (8 x 10")
Catalog # 12985-074
Supplier:   VWR International
VWR® Talon® Support Stands, Cast Iron, Rectangular Base
Catalog # 12985-074
Supplier:  VWR International
VWR® Talon® Support Stands, Cast Iron, Rectangular Base
VWR® Talon® Support Stands, Cast Iron, Rectangular Base
20.3 x 25.4 cm (8 x 10")
Price
$3,695.98
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Support stand bases are constructed of cast iron with black enamel finish.
  • Cast iron construction with black enamel finish
  • Built-in support rod holder
  • Rectangular base
  • For use with VWR® Talon® lab frames
All bases feature a built-in support rod holder with locking knob that accepts a 13mm (1/2") diameter support rod (sold separately).
Documents
Especificaciones
  • Dimensions:
    20.3 x 25.4 cm (8 x 10")
Accessories
Información del producto
  • Catalog #
    Ø
    Material
    Availability
    Price
  • Support Rod
    Support Rod
    60079-535
    13 mm (1/2")
    Stainless steel
    Log in Para disponibilidad
    Each (1UN) - $859.15
Información del producto
  • Catalog #
    Ø
    Material
    Availability
    Price
  • Support Rod
    Support Rod
    60079-536
    13 mm (1/2")
    Stainless steel
    Log in Para disponibilidad
    Each (1UN) - $1,229.04