Orders LinkContactUsLinkComponent
VWR® Talon® Support Stands, Cast Iron, Rectangular Base
10.2 x 15.2 cm (4 x 6")
  12985-068
VWR® Talon® Support Stands, Cast Iron, Rectangular Base
  12985-068
VWR® Talon® Support Stands, Cast Iron, Rectangular Base
VWR® Talon® Support Stands, Cast Iron, Rectangular Base
10.2 x 15.2 cm (4 x 6")
$1,158.75
Each (1UN)
Support stand bases are constructed of cast iron with black enamel finish.
  • Cast iron construction with black enamel finish
  • Built-in support rod holder
  • Rectangular base
  • For use with VWR® Talon® lab frames
All bases feature a built-in support rod holder with locking knob that accepts a 13mm (1/2") diameter support rod (sold separately).
  • Dimensions:
    10.2 x 15.2 cm (4 x 6")
Accessories
  •  
    Description
    Ø
    Material
  • 60079-535
    Support Rod
    13 mm (1/2")
    Stainless steel
    Each (1UN) - $859.15
  •  
    Description
    Ø
    Material
  • 60079-536
    Support Rod
    13 mm (1/2")
    Stainless steel
    Each (1UN) - $1,229.04