Disintegration Testing, Quality Lab Accessories
Clear Glass Disintegration Beaker with Flat Top, 900 mL
Catalog # 11010-360
Supplier: QUALITY LAB ACCESSORIES
Número de proveedor #: GLA905-DB
Disintegration Testing, Quality Lab Accessories
Catalog # 11010-360
Supplier: QUALITY LAB ACCESSORIES
Jump to:
Product Details & Documents
QLA manufactures Disintegration Testing Accessories and Glassware for Agilent/VanKel disintegration testers.
- Fluted disks can be used with any brand of disintegration tester
All accessories are USP compliant and made in the USA.
Especificaciones
- Description:Clear Glass Disintegration Beaker with Flat Top, 900 mL