Orders LinkContactUsLinkComponent
Ward's® Lichen Slides
Supplier:   Avantor
Ward's® Lichen Slides
Supplier:  Avantor
Certificates
Ward's® Lichen Slides
Ward's® Lichen Slides
4 Options Available Below
Product Details & Documents
A composite of fungi and algae.
  • Ward's produced slides
  • Rigorous quality control standards
A great example of mutualism, lichen are a composite organism of fungi and algae. In these slides both parts of this relationship are easily seen.
Product Family Options
Información del producto
  • Orientation
    Stain
    Availability
    Price
  • 470176-858
    Ascocarps
    Ascocarps
    Section
    Quadruple
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $116.28
    $116.28
    Each (1UN)
       $116.28
  • 470181-106
    Crustose
    Crustose
    Section
    Quadruple
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $381.60
    $381.60
    Each (1UN)
       $381.60
  • Foliose
    Section
    Quadruple
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $127.70
    $127.70
    Each (1UN)
       $127.70
  • 470181-102
    Mycobiont
    Mycobiont
    Whole mount
    Fast Green and Mallory's
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $182.70
    $182.70
    Each (1UN)
       $182.70