Orders LinkContactUsLinkComponent
Detector Calibration Multi-Element Mix: Tb, U @ 5
Detector Calibration Multi-Element Mix: Tb, U @ 5
Detector Calibration Multi-Element Mix: Tb, U @ 5; Y, In, Ce, Tl @ 10; Co @ 20; Mg, Sc @ 25; 7Li @ 50; Be @ 100 µg/mL in 5% HNO3, 100 mL, Dilute HNO3
Catalog # 77863-153
Supplier:   LGC Standards
Número de proveedor #:  VHG-LDCAL-100
Detector Calibration Multi-Element Mix: Tb, U @ 5
Catalog # 77863-153
Supplier:  LGC Standards
CAS Number:  
Detector Calibration Multi-Element Mix: Tb, U @ 5
Detector Calibration Multi-Element Mix: Tb, U @ 5
Detector Calibration Multi-Element Mix: Tb, U @ 5; Y, In, Ce, Tl @ 10; Co @ 20; Mg, Sc @ 25; 7Li @ 50; Be @ 100 µg/mL in 5% HNO3, 100 mL, Dilute HNO3
CAS Number:  
Price
$3,880.00
Log in Para disponibilidad
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Especificaciones
Especificaciones
  • Matrix:
    Dilute HNO3
  • Description:
    Detector Calibration Multi-Element Mix: Tb, U @ 5; Y, In, Ce, Tl @ 10; Co @ 20; Mg, Sc @ 25; 7Li @ 50; Be @ 100 µg/mL in 5% HNO3
  • Size:
    100 mL
Especificaciones
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 