Detector Calibration Multi-Element Mix: Tb, U @ 5
Product Details & Documents
Especificaciones
Especificaciones
- Matrix:Dilute HNO3
- Description:Detector Calibration Multi-Element Mix: Tb, U @ 5; Y, In, Ce, Tl @ 10; Co @ 20; Mg, Sc @ 25; 7Li @ 50; Be @ 100 µg/mL in 5% HNO3
- Size:100 mL