Orders LinkContactUsLinkComponent
Double Polypropylene Bucket Systems, 5 Gallon, Contec®
Double Polypropylene Bucket Systems, 5 Gallon, Contec®
Double Polypropylene Bucket Systems, 5 Gallon, Contec®
Double Polypropylene Bucket Systems, 5 Gallon, Contec®
The Contec® Double Polypropylene Bucket System is a fully autoclavable stainless steel rolling cart with two polypropylene five gallon (20 l) buckets (red, gray), and stainless steel drip cap. The durable, yet lightweight construction makes the bucket system easy to maneuver around cleanrooms.
  • Compact, lightweight design makes the system ideal for smaller cleanrooms
  • Ideal for flat and curved Contec® mops
  • Compatible with most disinfectants
The system is available with either standard casters or heavy duty casters. The use of the Double Bucket System allows for color-coded differentiation between cleaning and waste solutions.
The five gallon (20 l) autoclavable buckets are compatible with two types of wringers, curved or flat, depending on the mop used.
Ordering information: Wringers are sold separately.
Product Family Options
  • Description
    Capacity
    Color
  • Double Polypropylene Bucket System with Standard Casters
    20 L (5 gal.)
    Grey/red
    Case (2UN)
    Case (2UN) - $20,449.57
    $20,449.57
    Case (2UN)
       $20,449.57
     : 2769-KIT
  • Double Polypropylene Bucket System with Heavy-Duty Casters
    20 L (5 gal.)
    Grey/red
    Case (2UN)
    Case (2UN) - $26,051.91
    $26,051.91
    Case (2UN)
       $26,051.91
     : 2770-KIT