Orders LinkContactUsLinkComponent
Stainless Steel Wringers, Contec®
Stainless Steel Wringers, Contec®
Supplier:   CONTEC, INC. MX
Stainless Steel Wringers, Contec®
Supplier:  CONTEC, INC. MX
Certificates
Stainless Steel Wringers, Contec®
Stainless Steel Wringers, Contec®
2 Options Available Below
Product Details & Documents
Variety of wringers designed for use with curved or flat mops.
  • Stainless steel for variety of Contec® bucket systems
  • Ideal for curved, flat, and string mops
  • Fully autoclavable
Contec® offers a variety of wringers available for use with curved mops (EasyCurve Mop) and flat mops (VertiKlean MAX Mop and QuickTask Mop). These wringers fit on 20 l bucket systems. The wringers are fully autovlavable and compatible with a wide variety of disinfectants.
Documents
Product Family Options
Información del producto
  • Capacity
    Availability
    Price
  • 76270-910
    Flat Wringer for Polypropylene Buckets
    Flat Wringer for Polypropylene Buckets
    20 L (5 gal.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $7,229.64
    $7,229.64
    Each (1UN)
       $7,229.64
    Supplier Number: 2691
  • 76270-912
    Curved Wringer for Polypropylene Buckets
    Curved Wringer for Polypropylene Buckets
    20 L (5 gal.)
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $7,879.56
    $7,879.56
    Each (1UN)
       $7,879.56
    Supplier Number: 2694