SP Bel-Art Trigger Sprayer, with 53 mm Adapter, Bel-Art Products, a part of SP
Catálogo # 89238-034
Proveedor: Bel-Art Products, a Part of SP
Especificaciones
- Descripción:Trigger Sprayer with Adapter
- Referencia:89238-034
- Nº de proveedor:116200050
Especificaciones
Acerca de este artículo
Add the convenience of adjustable spray to Scienceware® wash bottles.
- Polypropylene construction
- Chemically resistant
- Stainless steel spring-action pump resists corrosion
Trigger sprayers with adapters fit any Scienceware® bottle with an easy-to-fill, 53 mm wide closure.
The simple to install sprayer is perfect for OSHA compliant 16 oz. and 32 oz. Scienceware® non-vented, wide-mouth wash bottles. A stainless steel, spring-action pump in the trigger sprayer will not corrode with repeated exposure to 10% bleach disinfecting solution.
Información de pedido: The adapter plus trigger sprayer adds approximately 14 cm (51/2") height to a wash bottle. Sprayer without adapter fits most bottles with a 28 mm opening. They offer convenient, adjustable spray dispensing of a wide range of chemicals including bleach, ethanol, machine oil, and water.