Tweezers with Angulated Tips, Style 6, EXCELTA®
Catalog # 63042-760
Supplier: EXCELTA CORPORATION
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Descripción:Tweezers, flat sharp point, 3 star quality
- Matriz:Stainless steel 316L, anti-magnetic
- Curvature:70° angulated
- Referencia:63042-760
- Longitud total:112.5 mm (4.5")
- Nº de proveedor:6-SA
- Tip Width:0.007" (0.17 mm)
- Tip Length:0.1 mm (0.004")
Specifications
About this item
Style 6 tweezers are uniquely angled to get into difficult places. They feature strong precise tips.
- Uniquely angled for great accessability
- Excellent visibility
- Sharp pointed, exact and precise
The 'Style 6' Series tweezers feature 70° angled flat sharp precision pointed tips and can be utilized in a variety of applications, such as manipulating very minute components or parts. The angled tips allow for tweezing in restricted areas and enhance the visibility.We offer 3 different qualities. The higher the quality, the more precise the tips. They are offered in carbon steel (hard), stainless steel (rust-resistant) or stainless anti-magnetic steel (anti-magnetic).